siRNA Details
Gene Symbol: 3010026O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026543.3 | NP_080819.2 | 7 | 638 | ||
GenBank mRNA | AK003636.1 | - | 7 | 613 | ||
AK019392.1 | - | 7 | 639 | |||
BC028428.1 | - | 7 | 623 | 1315017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
