siRNA Details
Gene Symbol: Gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024216.1 | NP_077178.1 | 6 | 703 | ||
GenBank mRNA | AK016996.1 | - | 6 | 634 | ||
AK042405.1 | - | 1418 | ||||
AK080720.1 | - | 6 | 650 | |||
AK087968.1 | - | 6 | 654 | |||
AK146783.1 | - | 6 | 641 | |||
BC003341.1 | - | 6 | 703 | 3257412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
