siRNA Details
Gene Symbol: Dusp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024438.4 | NP_077758.1 | 3 | 544 | ||
GenBank mRNA | AB038769.1 | - | 3 | 499 | ||
AB051896.1 | - | 3 | 518 | |||
AF237618.1 | - | 3 | 716 | |||
AK010127.1 | - | 3 | 541 | |||
AK018369.1 | - | 3 | 201 | |||
AK077871.1 | - | 3 | 545 | |||
AK170170.1 | - | 3 | 766 | |||
BC021591.1 | - | 3 | 499 | 5149051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
