siRNA Details
Gene Symbol: Fahd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029629.2 | NP_083905.1 | 5 | 813 | ||
GenBank mRNA | AK002291.1 | - | 5 | 791 | ||
AK005134.1 | - | 4 | 1050 | |||
AK080868.1 | - | 5 | 799 | |||
AK170875.1 | - | 5 | 787 | |||
BC030182.1 | - | 5 | 764 | 5044017 |

More Information
Other Species Targeted

Gene Ontology Categories:
