siRNA Details
Gene Symbol: B230217C12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080935.1 | NP_001074404.1 | 2 | 377 | ||
GenBank mRNA | AK018341.1 | - | 2 | 300 | ||
AK031146.1 | - | 299 | ||||
AK035650.1 | - | 282 | ||||
AK042827.1 | - | 302 | ||||
AK080808.1 | - | 299 | ||||
AK081899.1 | - | 320 | ||||
AK082773.1 | - | 2 | 307 |
