siRNA Details
Gene Symbol: Fam120aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014080.1 | - | 2 | 288 | ||
AK021143.1 | - | 2 | 250 | |||
BC024672.1 | - | 383 | ||||
BC045201.1 | - | 2 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
