siRNA Details
Gene Symbol: Gtf2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167921.1 | NP_001161393.1 | 3 | 325 | ||
NM_001167922.1 | NP_001161394.1 | 3 | 427 | |||
NM_026584.3 | NP_080860.3 | 3 | 313 | |||
GenBank mRNA | AK021183.1 | - | 3 | 278 | ||
AK088342.1 | - | 3 | 338 | |||
AK146803.1 | - | 3 | 300 | |||
AK150415.1 | - | 3 | 271 | |||
AK153370.1 | - | 3 | 271 | |||
AK158783.1 | - | 3 | 407 | |||
BC020016.1 | - | 3 | 293 | 3501113 | ||
BY256665.1 | - | 3 | 314 | |||
CX239577.1 | - | 3 | 325 |

More Information
Other Species Targeted

Gene Ontology Categories:
