siRNA Details
Gene Symbol: Fdx1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039824.2 | NP_001034913.1 | 5 | 726 | ||
GenBank mRNA | AK008401.2 | - | 5 | 724 | ||
AK010211.1 | - | 5 | 697 | |||
AK020979.1 | - | 5 | 730 | |||
BC061189.1 | - | 4 | 813 |
Gene Symbol: Gm38431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004919.1 | - | 17 | 2930 | ||
AY275472.1 | - | 17 | 2916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
