siRNA Details
Gene Symbol: Rpl39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026594.2 | NP_080870.1 | 3 | 281 | ||
GenBank mRNA | AK005645.1 | - | 3 | 271 | ||
AK019681.1 | - | 3 | 283 | |||
BC023885.1 | - | 3 | 271 | 5151514 | ||
BC051469.1 | - | 3 | 216 | 1363159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
