siRNA Details
Gene Symbol: Hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026601.3 | NP_080877.3 | 3 | 497 | ||
GenBank mRNA | AA403499.1 | - | 4 | 506 | ||
BC024619.1 | - | 3 | 485 | 4923664 | ||
BC100401.1 | - | 3 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
