siRNA Details
Gene Symbol: Ndufb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026610.1 | NP_080886.1 | 1 | 142 | ||
GenBank mRNA | AK004958.1 | - | 1 | 140 | ||
AK009807.1 | - | 1 | 142 | |||
AK018707.1 | - | 1 | 142 | |||
BC056229.1 | - | 1 | 143 | 6475281 | ||
BC087907.1 | - | 1 | 140 | |||
BC096421.1 | - | 1 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
