siRNA Details
Gene Symbol: Ndufc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024220.2 | NP_077182.1 | 2 | 454 | ||
GenBank mRNA | AK002373.1 | - | 2 | 362 | ||
AK003380.2 | - | 2 | 357 | |||
AK008502.1 | - | 2 | 369 | |||
AK008889.1 | - | 2 | 362 | |||
AK019002.1 | - | 2 | 381 | |||
AK153547.1 | - | 2 | 384 | |||
AK166594.1 | - | 2 | 381 | |||
AK168342.1 | - | 2 | 406 | |||
BC002097.1 | - | 2 | 377 | 3497384 | ||
CK128102.1 | - | 2 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
