siRNA Details
Gene Symbol: Rnaseh2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026616.2 | NP_080892.1 | 4 | 586 | ||
GenBank mRNA | AF346604.1 | - | 4 | 584 | ||
AK011556.1 | - | 4 | 564 | |||
AK018776.1 | - | 4 | 575 | |||
BC024333.1 | - | 4 | 558 | 5040687 | ||
CD553809.1 | - | 1 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
