siRNA Details
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024221.3 | NP_077183.1 | 8, 9 | 846 | ||
GenBank mRNA | AK011810.1 | - | 810 | |||
AK014180.1 | - | 8, 9 | 801 | |||
AK014918.1 | - | 8, 9 | 845 | |||
AK153058.1 | - | 8, 9 | 800 | |||
AK166631.1 | - | 8, 9 | 848 | |||
AK167798.1 | - | 8, 9 | 802 | |||
BC002188.1 | - | 6, 7 | 670 | |||
BC019512.1 | - | 8, 9 | 819 | 4210204 | ||
BC094468.1 | - | 8, 9 | 792 | 5011266 |

More Information
Other Species Targeted

Gene Ontology Categories:
