siRNA Details
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024221.3 | NP_077183.1 | 7 | 711 | ||
GenBank mRNA | AK011810.1 | - | 675 | |||
AK014180.1 | - | 7 | 666 | |||
AK014918.1 | - | 7 | 712 | |||
AK153058.1 | - | 7 | 665 | |||
AK166631.1 | - | 7 | 713 | |||
AK167798.1 | - | 7 | 667 | |||
AK218654.1 | - | 2 | 250 | |||
BC002188.1 | - | 5 | 535 | |||
BC019512.1 | - | 7 | 684 | 4210204 | ||
BC094468.1 | - | 7 | 657 | 5011266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
