siRNA Details
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177576.1 | NP_001171047.1 | 5 | 483 | ||
NM_026646.3 | NP_080922.1 | 5 | 725 | |||
GenBank mRNA | AK010193.1 | - | 5 | 471 | ||
AK018760.1 | - | 5 | 489 | |||
AK046874.1 | - | 5 | 470 | |||
AK048389.1 | - | 5 | 454 | |||
AK049313.1 | - | 5 | 454 | |||
AK155276.1 | - | 5 | 477 | |||
AK158985.1 | - | 1 | 2652 | |||
AK162400.1 | - | 5 | 436 | |||
BC037949.1 | - | 5 | 482 | 5367108 | ||
BC050887.1 | - | 5 | 678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
