siRNA Details
Gene Symbol: Zdhhc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026647.3 | NP_080923.2 | 5, 6 | 725 | ||
GenBank mRNA | AK020293.1 | - | 5, 6 | 607 | ||
AY166674.1 | - | 3, 4 | 363 | |||
BC043110.1 | - | 5, 6 | 571 | 6406930 | ||
BC058372.1 | - | 5, 6 | 593 |
