siRNA Details
Gene Symbol: Rbm28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156773.1 | - | 15 | 1599 | ||
BC018373.1 | - | 11 | 1036 | 3708613 | ||
BC027123.1 | - | 8 | 968 | 5356457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
