siRNA Details
Gene Symbol: Rbm28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156773.1 | - | 10 | 1102 | ||
BC018373.1 | - | 6 | 539 | 3708613 | ||
BC024602.1 | - | 10 | 1046 | |||
BC027123.1 | - | 3 | 471 | 5356457 | ||
BC040811.1 | - | 10 | 1110 | 4015830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
