siRNA Details
Gene Symbol: Rbm28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133925.2 | NP_598686.2 | 16 | 1838 | ||
GenBank mRNA | AK156773.1 | - | 16 | 1775 | ||
AK161670.1 | - | 16 | 1805 | |||
AK167050.1 | - | 16 | 1840 | |||
BC018373.1 | - | 12 | 1212 | 3708613 | ||
BC023293.1 | - | 6 | 587 | |||
BC024602.1 | - | 16 | 1719 | |||
BC027123.1 | - | 9 | 1144 | 5356457 | ||
BC040811.1 | - | 16 | 1783 | 4015830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
