siRNA Details
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005846.2 | NP_001005846.1 | 9 | 1115 | ||
NM_026656.4 | NP_080932.2 | 9 | 1143 | |||
GenBank mRNA | AF503575.1 | - | 9 | 1132 | ||
AK014467.1 | - | 9 | 1046 | |||
AK019454.1 | - | 9 | 1012 | |||
AK150446.1 | - | 9 | 1111 | |||
AK150547.1 | - | 9 | 1111 | |||
AK152067.1 | - | 9 | 1111 | |||
AK152943.1 | - | 9 | 1111 | |||
AK153290.1 | - | 9 | 1111 | |||
AK159172.1 | - | 9 | 1143 | |||
AY083532.1 | - | 8 | 961 | |||
BC029847.1 | - | 9 | 992 | 3967456 |

More Information
Other Species Targeted

Gene Ontology Categories:
