siRNA Details
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005846.2 | NP_001005846.1 | 5, 6 | 713 | ||
NM_026656.4 | NP_080932.2 | 5, 6 | 741 | |||
GenBank mRNA | AF503575.1 | - | 5, 6 | 730 | ||
AK014467.1 | - | 5, 6 | 644 | |||
AK019454.1 | - | 5, 6 | 610 | |||
AK150446.1 | - | 5, 6 | 709 | |||
AK150547.1 | - | 5, 6 | 709 | |||
AK152067.1 | - | 5, 6 | 709 | |||
AK152943.1 | - | 5, 6 | 709 | |||
AK153290.1 | - | 5, 6 | 709 | |||
AK159172.1 | - | 5, 6 | 741 | |||
AY083532.1 | - | 4, 5 | 559 | |||
BC029847.1 | - | 5, 6 | 590 | 3967456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
