siRNA Details
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005846.2 | NP_001005846.1 | 6, 7 | 810 | ||
NM_026656.4 | NP_080932.2 | 6, 7 | 838 | |||
GenBank mRNA | AF503575.1 | - | 6, 7 | 827 | ||
AK014467.1 | - | 6, 7 | 741 | |||
AK019454.1 | - | 6, 7 | 707 | |||
AK150446.1 | - | 6, 7 | 806 | |||
AK150547.1 | - | 6, 7 | 806 | |||
AK152067.1 | - | 6, 7 | 806 | |||
AK152943.1 | - | 6, 7 | 806 | |||
AK153290.1 | - | 6, 7 | 806 | |||
AK159172.1 | - | 6, 7 | 838 | |||
AY083532.1 | - | 5, 6 | 656 | |||
BC029847.1 | - | 6, 7 | 687 | 3967456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
