siRNA Details
Gene Symbol: Mfsd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026660.2 | NP_080936.1 | 8 | 1041 | ||
GenBank mRNA | AK003763.1 | - | 3 | 309 | ||
AK018736.1 | - | 8 | 1054 | |||
AK089055.1 | - | 8 | 1058 | |||
AK152438.1 | - | 8 | 1027 | |||
BC023012.1 | - | 8 | 1041 | 5376230 | ||
BC064775.1 | - | 8 | 939 | 30023877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
