siRNA Details
Gene Symbol: 0610011F06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026686.2 | NP_080962.1 | 6 | 704 | ||
GenBank mRNA | AK002535.1 | - | 6 | 690 | ||
AK151352.1 | - | 6 | 705 | |||
AK170337.1 | - | 6 | 700 | |||
BC069266.1 | - | 6 | 687 | 1248589 | ||
BX516844.1 | - | 1 | 169 |
