siRNA Details
Gene Symbol: Mul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026689.3 | NP_080965.2 | 1 | 155 | ||
GenBank mRNA | AK002416.1 | - | 1 | 137 | ||
AK044088.1 | - | 1 | 117 | |||
AK076419.1 | - | 1 | 115 | |||
AK083295.1 | - | 1 | 126 | |||
AK153956.1 | - | 1 | 105 | |||
AK170149.1 | - | 1 | 139 | |||
BB856128.1 | - | 1 | 157 | |||
BC019516.1 | - | 1 | 124 | 4163969 | ||
BY062056.1 | - | 1 | 146 | |||
EU935009.1 | - | 1 | 67 |

More Information
Other Species Targeted

Gene Ontology Categories:
