siRNA Details
Gene Symbol: 0610030E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026696.1 | NP_080972.1 | 4 | 787 | ||
GenBank mRNA | AK002701.1 | - | 4 | 733 | ||
AK085662.1 | - | 1 | 1998 | |||
BC064080.1 | - | 4 | 770 | 1246433 | ||
BC066053.1 | - | 4 | 761 | 6827710 | ||
BC117811.1 | - | 3 | 814 | |||
BC117812.1 | - | 4 | 737 | 40092656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
