siRNA Details
Gene Symbol: Mogat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026713.3 | NP_080989.2 | 6 | 859 | ||
GenBank mRNA | AF384162.1 | - | 5 | 736 | ||
AK002693.1 | - | 5 | 808 | |||
BC031920.1 | - | 883 | ||||
BC106135.1 | - | 5 | 807 | 5041577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
