siRNA Details
Gene Symbol: Nat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023455.3 | NP_075944.1 | 2 | 766 | ||
GenBank mRNA | AF163317.1 | - | 2 | 723 | ||
AK002784.1 | - | 2 | 707 | |||
BC019517.1 | - | 2 | 958 | 4236543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
