siRNA Details
Gene Symbol: G6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175935.3 | NP_787949.2 | 1 | 331 | ||
GenBank mRNA | AK002966.1 | - | 2 | 195 | ||
AK165395.1 | - | 1 | 185 | |||
AY186239.1 | - | 1 | 177 | |||
BC069959.1 | - | 1 | 331 | 4990136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
