siRNA Details
Gene Symbol: Lmbrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026719.2 | NP_080995.2 | 8 | 856 | ||
GenBank mRNA | AK032560.1 | - | 1155 | |||
AK152170.1 | - | 8 | 667 | |||
AK153237.1 | - | 8 | 667 | |||
AK158982.1 | - | 8 | 692 | |||
AK167080.1 | - | 8 | 675 | |||
AY061989.1 | - | 7 | 567 | |||
BC025579.1 | - | 8 | 688 | 5325113 | ||
BC027774.1 | - | 8 | 739 | 5346846 | ||
BC031902.1 | - | 8 | 679 | 5362166 | ||
BC039053.1 | - | 8 | 770 | 2655321 |
