siRNA Details
Gene Symbol: Ankrd13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026720.2 | NP_080996.2 | 4 | 534 | ||
GenBank mRNA | AK089186.1 | - | 4 | 527 | ||
AK155698.1 | - | 4 | 535 | |||
BC058982.1 | - | 4 | 445 | 5685059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
