siRNA Details
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290765.1 | NP_001277694.1 | 8 | 1037 | ||
NM_026721.3 | NP_080997.1 | 8 | 998 | |||
GenBank mRNA | AK003191.1 | - | 4 | 300 | ||
AK035296.1 | - | 8 | 905 | |||
BC100463.1 | - | 8 | 1005 | 30918840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
