siRNA Details
Gene Symbol: Steap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085409.1 | NP_001078878.1 | 4 | 1427 | ||
NM_133186.3 | NP_573449.2 | 3 | 1054 | |||
GenBank mRNA | AK003108.1 | - | 1028 | |||
AK028961.1 | - | 3 | 1031 | |||
AK078769.1 | - | 3 | 1448 | |||
AK167028.1 | - | 4 | 1427 | |||
AK171237.1 | - | 3 | 1059 | |||
AK217925.1 | - | 1 | 133 | |||
AY029586.1 | - | 3 | 1042 | |||
AY214462.1 | - | 1420 |
Gene Symbol: Steap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY029584.1 | - | 3 | 635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
