siRNA Details
Gene Symbol: Gpihbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026730.1 | NP_081006.1 | 3, 4 | 308 | ||
GenBank mRNA | AB095543.1 | - | 3, 4 | 315 | ||
AK003305.1 | - | 3, 4 | 324 | |||
BC061225.1 | - | 3, 4 | 308 | 30298145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
