siRNA Details
Gene Symbol: Ppp1r14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026731.3 | NP_081007.2 | 2 | 245 | ||
GenBank mRNA | AF352573.1 | - | 2 | 253 | ||
AK003249.1 | - | 2 | 244 | |||
AY050672.1 | - | 2 | 244 | |||
BC010832.1 | - | 2 | 245 | 4216018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
