siRNA Details
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 3 | 677 | ||
GenBank mRNA | AB041656.1 | - | 3 | 671 | ||
AK003356.2 | - | 3 | 678 | |||
AK004482.1 | - | 1 | 556 | |||
AK088418.1 | - | 3 | 609 | |||
AK158412.1 | - | 794 | ||||
AK183655.1 | - | 1 | 226 | |||
BC023033.1 | - | 3 | 640 | |||
BC037081.1 | - | 3 | 654 | 5098653 | ||
BC048360.1 | - | 3 | 656 | 5067090 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
