siRNA Details
Gene Symbol: 1110007C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026738.2 | NP_081014.1 | 2 | 180 | ||
GenBank mRNA | AK003526.1 | - | 2 | 181 | ||
AK154517.1 | - | 2 | 152 | |||
BC023485.1 | - | 3 | 312 | |||
BC047205.1 | - | 2 | 220 | |||
BC147073.1 | - | 2 | 136 | 9056051 | ||
BC147074.1 | - | 2 | 136 | 9056042 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134572, XM_008771547, XM_006253730, XM_008771548.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
