siRNA Details
Gene Symbol: Ndufaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026742.4 | NP_081018.1 | 3 | 421 | ||
GenBank mRNA | AK003553.2 | - | 3 | 376 | ||
AK034728.1 | - | 3 | 404 | |||
AK160341.1 | - | 3 | 417 | |||
BC028764.1 | - | 3 | 390 | 1195846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
