siRNA Details
Gene Symbol: Mrpl53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026744.3 | NP_081020.1 | 1 | 47 | ||
GenBank mRNA | AK003549.1 | - | 1 | 48 | ||
BC022162.1 | - | 1 | 72 | 5148541 | ||
BC065995.1 | - | 1270 | 5688315 |

More Information
Other Species Targeted

Gene Ontology Categories:
