siRNA Details
Gene Symbol: Nsmce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026746.3 | NP_081022.2 | 4, 5 | 591 | ||
GenBank mRNA | AK003685.1 | - | 4, 5 | 543 | ||
BC009125.1 | - | 4, 5 | 502 | 3155917 | ||
DV057668.1 | - | 4, 5 | 611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
