siRNA Details
Gene Symbol: Vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081041.1 | NP_001074510.1 | 1 | 217 | ||
GenBank mRNA | AK014229.1 | - | 1 | 231 | ||
AK137155.1 | - | 1 | 227 | |||
AK146211.1 | - | 1 | 226 | |||
BC034452.1 | - | 1 | 233 | |||
BC051479.1 | - | 1 | 223 | |||
BC118930.2 | - | 1 | 232 | 40090408 | ||
BC118931.1 | - | 1 | 232 | 40090409 |

More Information
Other Species Targeted

Gene Ontology Categories:
