siRNA Details
Gene Symbol: Vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081041.1 | NP_001074510.1 | 1 | 191 | ||
GenBank mRNA | AK014229.1 | - | 1 | 205 | ||
AK137155.1 | - | 1 | 201 | |||
AK146211.1 | - | 1 | 200 | |||
BC034452.1 | - | 1 | 207 | |||
BC051479.1 | - | 1 | 197 | |||
BC118930.2 | - | 1 | 206 | 40090408 | ||
BC118931.1 | - | 1 | 206 | 40090409 |
