siRNA Details
Gene Symbol: Ptx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163416.1 | NP_001156888.1 | 3 | 974 | ||
NM_026747.1 | NP_081023.1 | 3 | 1064 | |||
GenBank mRNA | AK003777.1 | - | 2 | 533 | ||
BC107380.2 | - | 3 | 957 | 40057084 | ||
BC107381.2 | - | 3 | 1047 | 40057088 | ||
BC115902.1 | - | 3 | 602 | |||
BC118508.1 | - | 3 | 1047 | 40106397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
