siRNA Details
Gene Symbol: Tomm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099675.1 | NP_001093145.1 | 2 | 354 | ||
NM_001134646.1 | NP_001128118.1 | 3 | 404 | |||
GenBank mRNA | AA423234.1 | - | 2 | 354 | ||
AK003822.1 | - | 2 | 350 | |||
CA466100.1 | - | 3 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
