siRNA Details
Gene Symbol: Zfyve21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026752.4 | NP_081028.3 | 4 | 440 | ||
GenBank mRNA | AK003661.1 | - | 4 | 337 | ||
AK139356.1 | - | 375 | ||||
AK155300.1 | - | 4 | 441 | |||
AK171099.1 | - | 4 | 381 | |||
AK192012.1 | - | 2 | 51 | |||
AK197357.1 | - | 2 | 51 | |||
AK216869.1 | - | 2 | 172 | |||
BC019521.1 | - | 4 | 370 | 4220205 | ||
BC132248.1 | - | 4 | 340 | 40130525 | ||
BC132250.1 | - | 4 | 340 | 40130527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
