siRNA Details
Gene Symbol: Tmem109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134142.1 | NP_598903.1 | 2 | 257 | ||
GenBank mRNA | AK049148.1 | - | 2 | 261 | ||
AK150779.1 | - | 2 | 241 | |||
AK151850.1 | - | 2 | 241 | |||
AK152419.1 | - | 2 | 241 | |||
AK165990.1 | - | 2 | 215 | |||
BC022997.1 | - | 2 | 257 | 5362560 | ||
BC023901.1 | - | 2 | 215 | 5321274 | ||
BC025033.1 | - | 2 | 227 | 4947460 | ||
BC025815.1 | - | 2 | 249 | 5097938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
