siRNA Details
Gene Symbol: Caly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190385.1 | NP_001177314.1 | 4 | 377 | ||
NM_001190386.1 | NP_001177315.1 | 5 | 657 | |||
NM_026769.5 | NP_081045.2 | 4 | 377 | |||
GenBank mRNA | AK002979.1 | - | 4 | 338 | ||
AK003400.2 | - | 4 | 379 | |||
BC049663.1 | - | 5 | 624 | 6532801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
