siRNA Details
Gene Symbol: Lamtor5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026774.2 | NP_081050.2 | 3 | 420 | ||
GenBank mRNA | AI552736.1 | - | 3 | 420 | ||
AK003358.1 | - | 3 | 260 | |||
AK146289.1 | - | 3 | 280 | |||
BC028547.1 | - | 3 | 250 | 1513442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
