siRNA Details
Gene Symbol: Dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172400.3 | NP_765988.2 | 2 | 173 | ||
GenBank mRNA | AK003892.2 | - | 2 | 163 | ||
AK049526.1 | - | 2 | 166 | |||
AK088325.1 | - | 2 | 177 | |||
AK150933.1 | - | 2 | 168 | |||
AK168645.1 | - | 2 | 157 | |||
BC035539.1 | - | 2 | 132 | |||
BC057191.1 | - | 2 | 144 | |||
BC064732.1 | - | 2 | 152 | |||
BC066222.1 | - | 2 | 152 | 30459247 | ||
BC080760.1 | - | 2 | 152 | |||
BC137735.1 | - | 2 | 169 | 8860754 | ||
BC145655.1 | - | 2 | 169 | |||
BY754536.1 | - | 2 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
